siRNA Details
| Gene Symbol: EIF3J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284335.1 | NP_001271264.1 | 3 | 318 | ||
| NM_003758.3 | NP_003749.2 | 3 | 318 | |||
| GenBank mRNA | AK002006.1 | - | 3 | 238 | ||
| AK023388.1 | - | 3 | 268 | |||
| AK300659.1 | - | 3 | 203 | |||
| BC002719.2 | - | 3 | 251 | 3632801 | ||
| U97670.1 | - | 3 | 251 | |||

EIF3J
Genomic Map