siRNA Details
| Gene Symbol: SEPHS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012248.3 | NP_036380.2 | 1 | 910 | ||
| GenBank mRNA | AK092003.1 | - | 3 | 662 | ||
| AK093889.1 | - | 2 | 667 | |||
| AK303275.1 | - | 2 | 722 | |||
| BC002381.2 | - | 1 | 794 | 2961275 | ||
| BC016643.1 | - | 1 | 783 | 3923979 | ||
| U43286.1 | - | 1 | 800 | |||

SEPHS2
Genomic Map