siRNA Details
| Gene Symbol: FBXL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278317.1 | NP_001265246.1 | 4 | 1796 | ||
| NM_012304.4 | NP_036436.1 | 4 | 2028 | |||
| XM_011513998.1 | XP_011512300.1 | 4 | 1587 | |||
| XM_017009262.1 | XP_016864751.1 | 4 | 6008 | |||
| GenBank mRNA | AB020647.2 | - | 4 | 2028 | ||
| BC136424.1 | - | 4 | 1793 | 9020413 | ||

FBXL7
Genomic Map