siRNA Details
| Gene Symbol: FBXL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278317.1 | NP_001265246.1 | 4 | 4166 | ||
| NM_012304.4 | NP_036436.1 | 4 | 4398 | |||
| XM_011513998.1 | XP_011512300.1 | 4 | 3957 | |||
| XM_017009262.1 | XP_016864751.1 | 4 | 8378 | |||
| GenBank mRNA | AB020647.2 | - | 4 | 4398 | ||
| AK021671.1 | - | 1 | 1366 | |||
| BC050571.1 | - | 1 | 1983 | |||

FBXL7
Genomic Map