siRNA Details
| Gene Symbol: KIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012311.3 | NP_036443.1 | 5, 6 | 660 | ||
| NR_045609.1 | 5, 6 | 660 | ||||
| XM_006717434.3 | XP_006717497.1 | 5, 6 | 655 | |||
| XM_006717435.3 | XP_006717498.1 | 4, 5 | 528 | |||
| XM_011519427.1 | XP_011517729.1 | 5, 6 | 655 | |||
| XM_017015946.1 | XP_016871435.1 | 4, 5 | 531 | |||
| GenBank mRNA | AJ005273.1 | - | 5, 6 | 631 | ||
| AK296637.1 | - | 4, 5 | 528 | |||
| AK301789.1 | - | 5, 6 | 660 | |||
| AK309059.1 | - | 5, 6 | 596 | |||
| BC017309.2 | - | 5, 6 | 577 | 5089578 | ||

KIN
Genomic Map