siRNA Details
| Gene Symbol: WBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012478.3 | NP_036610.2 | 2 | 212 | ||
| XM_011524580.2 | XP_011522882.1 | 2 | 133 | |||
| GenBank mRNA | AF087866.1 | - | 2 | 116 | ||
| AK057881.1 | - | 2 | 117 | |||
| AK128407.1 | - | 1 | 704 | |||
| AK293214.1 | - | 3 | 266 | |||
| AK294082.1 | - | 2 | 127 | |||
| AK294155.1 | - | 2 | 127 | |||
| AK300875.1 | - | 2 | 127 | |||
| BC007452.1 | - | 2 | 115 | 3140484 | ||
| BC010616.1 | - | 2 | 125 | 4177319 | ||
| DA278794.1 | - | 2 | 212 | |||

WBP2
Genomic Map