siRNA Details
| Gene Symbol: HABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014282.3 | NP_055097.2 | 7 | 1153 | ||
| XM_005251812.3 | XP_005251869.1 | 7 | 984 | |||
| XM_017014484.1 | XP_016869973.1 | 7 | 815 | |||
| GenBank mRNA | AF241831.1 | - | 7 | 1103 | ||
| AK000610.1 | - | 2 | 194 | |||
| AK025144.1 | - | 5 | 500 | |||
| AK055161.1 | - | 2 | 1511 | |||
| AK091242.1 | - | 2 | 2386 | |||
| AK124836.1 | - | 1 | 2261 | |||
| BC018788.1 | - | 4 | 792 | 3502216 | ||
| U77327.1 | - | 6 | 686 | |||

HABP4
Genomic Map