siRNA Details
| Gene Symbol: FAM120A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286722.1 | NP_001273651.1 | 18 | 4355 | ||
| NM_014612.4 | NP_055427.2 | 18 | 4358 | |||
| XM_005251842.4 | XP_005251899.1 | 18 | 4307 | |||
| XM_011518412.2 | XP_011516714.1 | 19 | 4259 | |||
| XM_011518413.2 | XP_011516715.1 | 19 | 4256 | |||
| XM_011518414.2 | XP_011516716.1 | 19 | 4256 | |||
| GenBank mRNA | BC098584.1 | - | 18 | 4334 | 30531603 | |
| BQ421167.1 | - | 1 | 714 | |||
| BX326996.1 | - | 1 | 514 | |||
| D80005.2 | - | 18 | 4118 | |||

FAM120A
Genomic Map