siRNA Details
| Gene Symbol: COBLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278458.1 | NP_001265387.1 | 13, 14 | 1895 | ||
| NM_001278460.1 | NP_001265389.1 | 10, 11 | 1718 | |||
| NM_001278461.1 | NP_001265390.1 | 9, 10 | 1678 | |||
| NM_014900.4 | NP_055715.3 | 10, 11 | 1795 | |||
| GenBank mRNA | AB023194.1 | - | 10, 11 | 1789 | ||
| AK225849.1 | - | 10, 11 | 1575 | |||
| AK294937.1 | - | 13, 14 | 1895 | |||
| BC071588.1 | - | 9, 10 | 1680 | 30345650 | ||
| BX537877.1 | - | 5, 6 | 3280 | |||
| BX649112.1 | - | 4, 5 | 661 | |||

COBLL1
Genomic Map