siRNA Details
| Gene Symbol: FRMPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014907.2 | NP_055722.2 | 4 | 395 | ||
| XM_011517803.2 | XP_011516105.1 | 5 | 797 | |||
| XM_011517804.2 | XP_011516106.1 | 4 | 716 | |||
| XM_017014480.1 | XP_016869969.1 | 4 | 1076 | |||
| GenBank mRNA | AB023184.1 | - | 4 | 395 | ||
| BC114965.1 | - | 4 | 374 | 40082639 | ||
| BX648967.1 | - | 4 | 945 | |||

FRMPD1
Genomic Map