siRNA Details
| Gene Symbol: ADGRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008701.2 | NP_001008701.1 | 10 | 2128 | ||
| NM_014921.4 | NP_055736.2 | 9 | 2113 | |||
| XM_005259818.3 | XP_005259875.1 | 9 | 2169 | |||
| XM_011527796.2 | XP_011526098.1 | 10 | 2205 | |||
| XM_011527798.2 | XP_011526100.1 | 9 | 2190 | |||
| XM_011527801.2 | XP_011526103.1 | 8 | 1645 | |||
| XM_017026475.1 | XP_016881964.1 | 10 | 2208 | |||
| XM_017026476.1 | XP_016881965.1 | 11 | 2318 | |||
| XM_017026477.1 | XP_016881966.1 | 10 | 2187 | |||
| XM_017026478.1 | XP_016881967.1 | 10 | 2208 | |||
| XM_017026479.1 | XP_016881968.1 | 10 | 2208 | |||
| XM_017026480.1 | XP_016881969.1 | 7 | 1623 | |||
| XM_017026481.1 | XP_016881970.1 | 10 | 2212 | |||
| GenBank mRNA | AB020628.1 | - | 10 | 2130 | ||
| AF307079.1 | - | 9 | 2097 | |||
More Information
Set Membership: |
|

ADGRL1
Genomic Map