siRNA Details
| Gene Symbol: RPH3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143854.1 | NP_001137326.1 | 10 | 1141 | ||
| NM_014954.3 | NP_055769.2 | 9 | 1129 | |||
| XM_011538048.1 | XP_011536350.1 | 10 | 945 | |||
| GenBank mRNA | AB023202.1 | - | 10 | 1062 | ||
| AK295696.1 | - | 10 | 1095 | |||
| AK296296.1 | - | 8 | 680 | |||
| AK316112.1 | - | 10 | 976 | |||
| BC002605.2 | - | 8 | 867 | |||
| BC009843.2 | - | 8 | 790 | |||
| BC017259.2 | - | 9 | 936 | 3510158 | ||
| BX647281.1 | - | 2 | 1735 | |||

RPH3A
Genomic Map