siRNA Details
| Gene Symbol: BTBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282550.1 | NP_001269479.1 | 5 | 1344 | ||
| NM_001282551.1 | NP_001269480.1 | 5 | 1440 | |||
| NM_001282552.2 | NP_001269481.1 | 4 | 1434 | |||
| NM_001282554.1 | NP_001269483.1 | 4 | 1349 | |||
| NM_014962.3 | NP_055777.1 | 4 | 1762 | |||
| NM_181443.3 | NP_852108.2 | 5 | 1525 | |||
| XM_017027728.1 | XP_016883217.1 | 6 | 1673 | |||
| XM_017027729.1 | XP_016883218.1 | 6 | 1605 | |||
| XM_017027730.1 | XP_016883219.1 | 3 | 1693 | |||
| GenBank mRNA | AB023169.1 | - | 4 | 1710 | ||
| AK296364.1 | - | 3 | 988 | |||
| AK299281.1 | - | 5 | 1525 | |||
| AK300371.1 | - | 3 | 1045 | |||
| AK307472.1 | - | 4 | 1092 | |||
| AK316256.1 | - | 4 | 1325 | |||
| AK316519.1 | - | 3 | 1268 | |||
| AL137756.1 | - | 1 | 2514 | |||
| BC093677.1 | - | 4 | 1233 | |||
| BC101527.1 | - | 4 | 1232 | 8069033 | ||
| BC109315.1 | - | 5 | 1373 | 40036410 | ||
| BC109316.1 | - | 5 | 1373 | 40036411 | ||

BTBD3
Genomic Map