siRNA Details
| Gene Symbol: SNX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003794.3 | NP_003785.1 | 4 | 508 | ||
| NR_073435.1 | 3 | 386 | ||||
| XM_017007414.1 | XP_016862903.1 | 4 | 500 | |||
| GenBank mRNA | AF065485.1 | - | 4 | 466 | ||
| AK001835.1 | - | 4 | 470 | |||
| AK298972.1 | - | 3 | 353 | |||
| BC018762.1 | - | 4 | 484 | 4641014 | ||
| BI669801.1 | - | 4 | 512 | |||
| DA289870.1 | - | 4 | 510 | |||

SNX4
Genomic Map