siRNA Details
| Gene Symbol: WDTC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276252.1 | NP_001263181.1 | 16 | 4403 | ||
| NM_015023.4 | NP_055838.2 | 16 | 4400 | |||
| XM_011541057.1 | XP_011539359.1 | 16 | 4081 | |||
| GenBank mRNA | AB028960.1 | - | 10 | 4746 | ||
| AK001734.1 | - | 16 | 3091 | |||
| AK023101.1 | - | 7 | 2942 | |||
| AK023778.1 | - | 7 | 2982 | |||
| BC032523.1 | - | 16 | 4263 | 5500384 | ||

WDTC1
Genomic Map