siRNA Details
| Gene Symbol: XPO7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015024.4 | NP_055839.3 | 2 | 207 | ||
| XM_017013244.1 | XP_016868733.1 | 2 | 434 | |||
| XM_017013245.1 | XP_016868734.1 | 2 | 201 | |||
| XM_017013246.1 | XP_016868735.1 | 2 | 430 | |||
| XM_017013247.1 | XP_016868736.1 | 2 | 594 | |||
| GenBank mRNA | AB018288.1 | - | 2 | 117 | ||
| AF064729.1 | - | 2 | 117 | |||
| AK291472.1 | - | 2 | 122 | |||
| AK311616.1 | - | 2 | 300 | |||
| AL597247.1 | - | 2 | 163 | |||
| BC030785.1 | - | 2 | 206 | 5269230 | ||
| DB460798.1 | - | 2 | 209 | |||
| DC332379.1 | - | 2 | 129 | |||

XPO7
Genomic Map