siRNA Details
| Gene Symbol: GGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015044.4 | NP_055859.1 | 4 | 391 | ||
| XM_017023074.1 | XP_016878563.1 | 5 | 4080 | |||
| XM_017023075.1 | XP_016878564.1 | 4 | 394 | |||
| GenBank mRNA | AB029003.1 | - | 1 | 21 | ||
| AF165531.1 | - | 4 | 329 | |||
| AF190863.1 | - | 4 | 335 | |||
| AF233522.1 | - | 4 | 334 | |||
| AF323754.1 | - | 4 | 333 | |||
| AK024619.1 | - | 4 | 390 | |||
| AK308425.1 | - | 4 | 378 | |||
| BC000284.1 | - | 4 | 373 | 3356490 | ||
| DA439738.1 | - | 4 | 391 | |||

GGA2
Genomic Map