siRNA Details
| Gene Symbol: ZC3H13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015070.4 | NP_055885.3 | 17 | 5471 | ||
| XM_005266312.1 | XP_005266369.1 | 17 | 5765 | |||
| GenBank mRNA | AL136745.1 | - | 10 | 4507 | ||
| AY283618.1 | - | 17 | 5469 | |||

ZC3H13
Genomic Map