siRNA Details
| Gene Symbol: SPG20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142294.1 | NP_001135766.1 | 2 | 486 | ||
| NM_001142295.1 | NP_001135767.1 | 2 | 610 | |||
| NM_001142296.1 | NP_001135768.1 | 3 | 662 | |||
| NM_015087.4 | NP_055902.1 | 2 | 679 | |||
| XM_005266313.4 | XP_005266370.1 | 2 | 604 | |||
| XM_005266314.3 | XP_005266371.1 | 3 | 653 | |||
| XM_005266315.3 | XP_005266372.1 | 3 | 507 | |||
| XM_005266317.3 | XP_005266374.1 | 2 | 437 | |||
| XM_011535012.2 | XP_011533314.1 | 2 | 590 | |||
| GenBank mRNA | AB011182.1 | - | 2 | 453 | ||
| AK001949.1 | - | 2 | 443 | |||
| AK291724.1 | - | 2 | 530 | |||
| AY038359.1 | - | 1 | 392 | |||
| AY123329.1 | - | 2 | 453 | |||
| BC047083.1 | - | 2 | 486 | 5313378 | ||
| BX648004.1 | - | 2 | 575 | |||
| KU178563.1 | - | 1 | 392 | |||
| KU178564.1 | - | 1 | 392 | |||

SPG20
Genomic Map