siRNA Details
| Gene Symbol: SPG20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142294.1 | NP_001135766.1 | 9 | 4434 | ||
| NM_001142295.1 | NP_001135767.1 | 9 | 4558 | |||
| NM_001142296.1 | NP_001135768.1 | 10 | 4610 | |||
| NM_015087.4 | NP_055902.1 | 9 | 4627 | |||
| XM_005266313.4 | XP_005266370.1 | 9 | 4552 | |||
| XM_005266314.3 | XP_005266371.1 | 10 | 4601 | |||
| XM_005266315.3 | XP_005266372.1 | 10 | 4455 | |||
| XM_005266317.3 | XP_005266374.1 | 9 | 4385 | |||
| XM_011535012.2 | XP_011533314.1 | 9 | 4538 | |||

SPG20
Genomic Map