siRNA Details
| Gene Symbol: CIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304815.1 | NP_001291744.1 | 11 | 4980 | ||
| NM_015125.4 | NP_055940.3 | 10 | 2225 | |||
| XM_005258673.2 | XP_005258730.1 | 10 | 2317 | |||
| XM_005258674.2 | XP_005258731.1 | 10 | 2317 | |||
| XM_005258675.2 | XP_005258732.1 | 10 | 2317 | |||
| XM_011526660.2 | XP_011524962.1 | 11 | 5022 | |||
| XM_011526661.2 | XP_011524963.1 | 11 | 5022 | |||
| XM_011526662.2 | XP_011524964.1 | 11 | 5022 | |||
| XM_011526663.2 | XP_011524965.1 | 11 | 5022 | |||
| XM_011526664.2 | XP_011524966.1 | 11 | 5022 | |||
| XM_011526665.2 | XP_011524967.1 | 11 | 5022 | |||
| XM_011526666.2 | XP_011524968.1 | 11 | 5022 | |||
| XM_011526667.2 | XP_011524969.1 | 11 | 5022 | |||
| GenBank mRNA | AB002304.1 | - | 7 | 1716 | ||
| AF363689.1 | - | 10 | 2225 | |||

CIC
Genomic Map