siRNA Details
| Gene Symbol: SNX13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015132.4 | NP_055947.1 | 11 | 1247 | ||
| XM_005249672.1 | XP_005249729.1 | 11 | 1257 | |||
| XM_005249673.4 | XP_005249730.1 | 11 | 1257 | |||
| XM_011515229.1 | XP_011513531.1 | 11 | 1257 | |||
| XM_011515230.2 | XP_011513532.1 | 12 | 1304 | |||
| XM_011515231.2 | XP_011513533.1 | 13 | 1363 | |||
| XM_011515233.2 | XP_011513535.1 | 12 | 1305 | |||
| XM_017011893.1 | XP_016867382.1 | 11 | 1257 | |||
| XM_017011894.1 | XP_016867383.1 | 12 | 1704 | |||
| XM_017011895.1 | XP_016867384.1 | 12 | 1309 | |||
| XM_017011896.1 | XP_016867385.1 | 12 | 3217 | |||
| XM_017011897.1 | XP_016867386.1 | 11 | 1178 | |||
| GenBank mRNA | AB018256.1 | - | 11 | 1207 | ||
| AF420470.1 | - | 11 | 1215 | |||
| AK023740.1 | - | 6 | 492 | |||
| AK294582.1 | - | 13 | 1202 | |||
| AK315135.1 | - | 11 | 1233 | |||
| BC045667.1 | - | 11 | 1217 | 5298570 | ||

SNX13
Genomic Map