siRNA Details
| Gene Symbol: RCOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015156.3 | NP_055971.2 | 6 | 952 | ||
| GenBank mRNA | AF155595.1 | - | 6 | 715 | ||
| BC051003.1 | - | 6 | 950 | |||
| BC064495.1 | - | 6 | 950 | |||
| D31888.1 | - | 6 | 457 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RCOR1| Gene Symbol: RCOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015156.3 | NP_055971.2 | 6 | 952 | ||
| GenBank mRNA | AF155595.1 | - | 6 | 715 | ||
| BC051003.1 | - | 6 | 950 | |||
| BC064495.1 | - | 6 | 950 | |||
| D31888.1 | - | 6 | 457 | |||