siRNA Details
| Gene Symbol: ARC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015193.4 | NP_056008.1 | 3 | 2864 | ||
| GenBank mRNA | AF193421.1 | - | 3 | 2852 | ||
| BC012321.1 | - | 3 | 2868 | 4555433 | ||
| BC075802.1 | - | 3 | 2856 | 5198843 | ||
| D87468.1 | - | 3 | 2856 | |||

ARC
Genomic Map