siRNA Details
| Gene Symbol: CGGBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008390.1 | NP_001008391.1 | 4 | 2121 | ||
| NM_001195308.1 | NP_001182237.1 | 4 | 2102 | |||
| NM_003663.3 | NP_003654.3 | 3 | 2019 | |||
| XM_017007355.1 | XP_016862844.1 | 3 | 1927 | |||
| GenBank mRNA | AL117392.1 | - | 3 | 1962 | ||
| BC052980.1 | - | 3 | 1916 | 6012191 | ||

CGGBP1
Genomic Map