siRNA Details
| Gene Symbol: ADGRL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322246.1 | NP_001309175.1 | 26 | 6611 | ||
| NM_001322402.1 | NP_001309331.1 | 26 | 6763 | |||
| NM_015236.5 | NP_056051.2 | 25 | 6559 | |||
| XM_011531788.2 | XP_011530090.1 | 27 | 6319 | |||
| XM_011531791.2 | XP_011530093.1 | 24 | 6235 | |||
| XM_017007929.1 | XP_016863418.1 | 29 | 9221 | |||
| XM_017007930.1 | XP_016863419.1 | 28 | 8173 | |||
| XM_017007931.1 | XP_016863420.1 | 27 | 6430 | |||
| XM_017007932.1 | XP_016863421.1 | 26 | 6301 | |||
| XM_017007933.1 | XP_016863422.1 | 26 | 6292 | |||
| XM_017007934.1 | XP_016863423.1 | 26 | 6286 | |||
| XM_017007935.1 | XP_016863424.1 | 25 | 6274 | |||
| XM_017007936.1 | XP_016863425.1 | 25 | 6262 | |||
| XM_017007937.1 | XP_016863426.1 | 27 | 7969 | |||
| XM_017007938.1 | XP_016863427.1 | 24 | 6220 | |||
| XM_017007939.1 | XP_016863428.1 | 22 | 6016 | |||
| XM_017007940.1 | XP_016863429.1 | 27 | 6353 | |||
| XM_017007941.1 | XP_016863430.1 | 26 | 6326 | |||
| XM_017007942.1 | XP_016863431.1 | 26 | 6134 | |||
| GenBank mRNA | AB018311.1 | - | 13 | 4021 | ||
| AK000781.1 | - | 7 | 1802 | |||
More Information
Set Membership: |
|

ADGRL3
Genomic Map