siRNA Details
| Gene Symbol: ANKS1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015245.2 | NP_056060.2 | 4 | 597 | ||
| XM_005248964.3 | XP_005249021.1 | 5 | 648 | |||
| XM_006715036.2 | XP_006715099.1 | 5 | 648 | |||
| XM_011514431.2 | XP_011512733.1 | 5 | 648 | |||
| XM_011514432.2 | XP_011512734.1 | 5 | 648 | |||
| XM_011514433.1 | XP_011512735.1 | 5 | 648 | |||
| XM_011514434.2 | XP_011512736.1 | 4 | 585 | |||
| XM_011514435.2 | XP_011512737.1 | 4 | 585 | |||
| XM_011514436.2 | XP_011512738.1 | 5 | 4217 | |||
| GenBank mRNA | AK298987.1 | - | 4 | 555 | ||
| BC022396.1 | - | 4 | 597 | |||
| BC031934.1 | - | 5 | 669 | 4819948 | ||
| BC132832.1 | - | 4 | 534 | 40146854 | ||
| D86982.1 | - | 4 | 594 | |||

ANKS1A
Genomic Map