siRNA Details
| Gene Symbol: FCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK075140.1 | - | 7 | 667 | ||
| AY358081.1 | - | 7 | 656 | |||
| AY358283.1 | - | 7 | 693 | |||
| BC020731.1 | - | 7 | 667 | 4692272 | ||
| CR456808.1 | - | 8 | 694 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FCN3| Gene Symbol: FCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK075140.1 | - | 7 | 667 | ||
| AY358081.1 | - | 7 | 656 | |||
| AY358283.1 | - | 7 | 693 | |||
| BC020731.1 | - | 7 | 667 | 4692272 | ||
| CR456808.1 | - | 8 | 694 | |||