siRNA Details
| Gene Symbol: SASH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015278.3 | NP_056093.3 | 7 | 1047 | ||
| XM_011535640.1 | XP_011533942.1 | 7 | 490 | |||
| XM_017010598.1 | XP_016866087.1 | 7 | 1279 | |||
| XM_017010599.1 | XP_016866088.1 | 7 | 602 | |||
| XM_017010600.1 | XP_016866089.1 | 7 | 490 | |||
| XM_017010601.1 | XP_016866090.1 | 6 | 704 | |||
| XM_017010602.1 | XP_016866091.1 | 5 | 401 | |||
| XM_017010603.1 | XP_016866092.1 | 5 | 340 | |||
| XM_017010604.1 | XP_016866093.1 | 4 | 249 | |||
| XM_017010605.1 | XP_016866094.1 | 4 | 215 | |||
| XM_017010607.1 | XP_016866096.1 | 8 | 670 | |||
| GenBank mRNA | AB018333.1 | - | 7 | 790 | ||
| AJ507735.1 | - | 7 | 578 | |||
| AK024403.1 | - | 1 | 32 | |||
| AK025495.1 | - | 7 | 1047 | |||
| BC063279.1 | - | 7 | 516 | |||

SASH1
Genomic Map