siRNA Details
| Gene Symbol: LEPROTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015344.2 | NP_056159.2 | 3, 4 | 386 | ||
| GenBank mRNA | AF063605.1 | - | 3, 4 | 363 | ||
| AF161461.1 | - | 3, 4 | 319 | |||
| AK075363.1 | - | 3, 4 | 330 | |||
| AY277595.1 | - | 3, 4 | 358 | |||
| AY358674.1 | - | 3, 4 | 358 | |||
| BC000642.2 | - | 3, 4 | 369 | 3344487 | ||
| CR541735.1 | - | 2, 3 | 279 | |||
| CR541760.1 | - | 2, 3 | 279 | |||
| DA258969.1 | - | 3, 4 | 387 | |||

LEPROTL1
Genomic Map