siRNA Details
| Gene Symbol: LEPROTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015344.2 | NP_056159.2 | 4 | 2210 | ||
| GenBank mRNA | AF063605.1 | - | 4 | 2187 | ||
| AK075363.1 | - | 4 | 2155 | |||
| AY358674.1 | - | 4 | 2182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LEPROTL1| Gene Symbol: LEPROTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015344.2 | NP_056159.2 | 4 | 2210 | ||
| GenBank mRNA | AF063605.1 | - | 4 | 2187 | ||
| AK075363.1 | - | 4 | 2155 | |||
| AY358674.1 | - | 4 | 2182 | |||