siRNA Details
| Gene Symbol: SAMD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161576.2 | NP_001155048.2 | 6 | 1618 | ||
| NM_001161577.1 | NP_001155049.1 | 3 | 511 | |||
| NM_015589.5 | NP_056404.4 | 7 | 1882 | |||
| GenBank mRNA | AB028976.1 | - | 5 | 949 | ||
| AK024652.1 | - | 7 | 1575 | |||
| AK025923.1 | - | 5 | 773 | |||
| BC021060.1 | - | 3 | 428 | |||
| BC021726.2 | - | 3 | 466 | |||
| BC121173.1 | - | 6 | 1315 | 40122465 | ||
| BC121174.1 | - | 6 | 1315 | 40122467 | ||
| BI825501.1 | - | 3 | 511 | |||

SAMD4A
Genomic Map