siRNA Details
| Gene Symbol: SAMD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161576.2 | NP_001155048.2 | 10 | 2112 | ||
| NM_001161577.1 | NP_001155049.1 | 8 | 1113 | |||
| NM_015589.5 | NP_056404.4 | 11 | 2376 | |||
| GenBank mRNA | AB028976.1 | - | 9 | 1443 | ||
| AK024652.1 | - | 11 | 2069 | |||
| AK025923.1 | - | 9 | 1257 | |||
| AL117523.1 | - | 3 | 523 | |||
| BC021060.1 | - | 7 | 922 | |||
| BC021726.2 | - | 8 | 1068 | |||
| BC121173.1 | - | 10 | 1809 | 40122465 | ||
| BC121174.1 | - | 10 | 1809 | 40122467 | ||

SAMD4A
Genomic Map