siRNA Details
| Gene Symbol: GNB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006578.3 | NP_006569.1 | 10 | 1072 | ||
| NM_016194.3 | NP_057278.2 | 12 | 1177 | |||
| XM_011521162.2 | XP_011519464.1 | 10 | 1429 | |||
| XM_011521163.2 | XP_011519465.1 | 10 | 954 | |||
| XM_017021867.1 | XP_016877356.1 | 7 | 2695 | |||
| GenBank mRNA | AF017656.1 | - | 10 | 1085 | ||
| AF300650.1 | - | 11 | 1112 | |||
| AF501885.1 | - | 10 | 986 | |||
| AK094776.1 | - | 3 | 800 | |||
| AK125702.1 | - | 7 | 1593 | |||
| AK290330.1 | - | 10 | 1080 | |||
| AK314775.1 | - | 10 | 1030 | |||
| AL117471.1 | - | 8 | 784 | |||
| BC013997.2 | - | 10 | 1069 | 3621195 | ||

GNB5
Genomic Map