siRNA Details
| Gene Symbol: DLL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016941.3 | NP_058637.1 | 7, 8 | 1739 | ||
| NM_203486.2 | NP_982353.1 | 7, 8 | 1739 | |||
| GenBank mRNA | AK075302.1 | - | 7, 8 | 1709 | ||
| BC000218.2 | - | 7, 8 | 1688 | 3508262 | ||
| BQ420593.1 | - | 1, 2 | 326 | |||

DLL3
Genomic Map