siRNA Details
| Gene Symbol: USP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017414.3 | NP_059110.2 | 6 | 865 | ||
| XM_006724074.2 | XP_006724137.1 | 6 | 936 | |||
| GenBank mRNA | AF176642.2 | - | 5 | 547 | ||
| AJ243526.1 | - | 6 | 730 | |||
| AK313385.1 | - | 6 | 686 | |||
| AL136690.1 | - | 6 | 686 | |||
| BC014896.1 | - | 6 | 671 | 3910720 | ||
| BT006835.1 | - | 5 | 527 | |||
| CR457216.1 | - | 5 | 527 | |||
| CT841507.1 | - | 5 | 593 | |||

USP18
Genomic Map