siRNA Details
| Gene Symbol: WDR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320546.1 | NP_001307475.1 | 2 | 1160 | ||
| NM_001320547.1 | NP_001307476.1 | 2 | 531 | |||
| NM_018031.4 | NP_060501.3 | 2 | 946 | |||
| GenBank mRNA | AF099100.1 | - | 2 | 615 | ||
| AK225653.1 | - | 2 | 609 | |||
| AK295145.1 | - | 2 | 946 | |||
| AK296387.1 | - | 2 | 231 | |||
| AK304117.1 | - | 2 | 531 | |||
| AK309913.1 | - | 2 | 826 | |||
| BC101707.1 | - | 2 | 605 | 8069213 | ||
| BC113467.1 | - | 2 | 605 | 8322519 | ||
| BC143486.1 | - | 2 | 605 | |||

WDR6
Genomic Map