siRNA Details
| Gene Symbol: SCARF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003693.3 | NP_003684.2 | 11 | 3296 | ||
| NM_145350.2 | NP_663325.1 | 11 | 3259 | |||
| NR_028075.2 | 11 | 3226 | ||||
| NR_102409.1 | 11 | 3333 | ||||
| XM_017025256.1 | XP_016880745.1 | 7 | 2197 | |||
| GenBank mRNA | AB052947.1 | - | 11 | 3157 | ||
| BC039735.1 | - | 11 | 3262 | 6041360 | ||
| D63483.1 | - | 11 | 3205 | |||
| D86864.1 | - | 11 | 3227 | |||

SCARF1
Genomic Map