siRNA Details
| Gene Symbol: SPACA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316897.1 | NP_001303826.1 | 3, 4 | 512 | ||
| NM_001316898.1 | NP_001303827.1 | 3, 4 | 505 | |||
| NM_001316900.1 | NP_001303829.1 | 3, 4 | 512 | |||
| NM_018956.4 | NP_061829.3 | 3, 4 | 512 | |||
| XM_017014231.1 | XP_016869720.1 | 3, 4 | 508 | |||
| GenBank mRNA | AJ011375.1 | - | 3, 4 | 500 | ||
| AK075259.1 | - | 3, 4 | 505 | |||
| BC012940.1 | - | 3, 4 | 462 | 4478288 | ||
| CK906175.1 | - | 3, 4 | 458 | |||
| CR457210.1 | - | 2, 3 | 351 | |||
| DA134573.1 | - | 3, 4 | 505 | |||

SPACA9
Genomic Map