siRNA Details
| Gene Symbol: ZNF273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021148.2 | NP_066971.2 | 4 | 3598 | ||
| NR_003099.1 | 5 | 3687 | ||||
| GenBank mRNA | AL832810.1 | - | 1 | 3290 | ||
| BC063818.1 | - | 5 | 3681 | 6503687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF273| Gene Symbol: ZNF273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021148.2 | NP_066971.2 | 4 | 3598 | ||
| NR_003099.1 | 5 | 3687 | ||||
| GenBank mRNA | AL832810.1 | - | 1 | 3290 | ||
| BC063818.1 | - | 5 | 3681 | 6503687 | ||