siRNA Details
| Gene Symbol: PLAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316887.1 | NP_001303816.1 | 4 | 909 | ||
| NM_001316888.1 | NP_001303817.1 | 3 | 937 | |||
| NM_001316889.1 | NP_001303818.1 | 2 | 819 | |||
| NM_021796.3 | NP_068568.1 | 3 | 891 | |||
| XM_011531257.2 | XP_011529559.1 | 3 | 817 | |||
| XM_011531260.2 | XP_011529562.1 | 3 | 931 | |||
| XM_017029236.1 | XP_016884725.1 | 2 | 859 | |||
| GenBank mRNA | AF234654.1 | - | 3 | 896 | ||
| AK075086.1 | - | 3 | 891 | |||
| BC022335.1 | - | 3 | 876 | 4769552 | ||
| BC066327.1 | - | 2 | 819 | 30339131 | ||

PLAC1
Genomic Map