siRNA Details
| Gene Symbol: ZNF277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021994.2 | NP_068834.2 | 2 | 383 | ||
| XM_005250129.3 | XP_005250186.1 | 2 | 274 | |||
| XM_011515768.2 | XP_011514070.1 | 2 | 355 | |||
| XM_017011720.1 | XP_016867209.1 | 2 | 266 | |||
| GenBank mRNA | AF209198.1 | - | 2 | 383 | ||
| AF308819.1 | - | 2 | 254 | |||
| AK027128.1 | - | 2 | 269 | |||
| AK314630.1 | - | 2 | 271 | |||
| AL713791.1 | - | 2 | 272 | |||
| BC020626.1 | - | 2 | 270 | 4616168 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172575, NM_178845, AK075945, AK156529, BY206876, and others.

ZNF277
Genomic Map