siRNA Details
| Gene Symbol: ZNF277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021994.2 | NP_068834.2 | 3 | 454 | ||
| XM_011515768.2 | XP_011514070.1 | 3 | 426 | |||
| GenBank mRNA | AF209198.1 | - | 3 | 454 | ||
| AF308819.1 | - | 3 | 325 | |||
| AK314630.1 | - | 3 | 342 | |||
| AL713791.1 | - | 3 | 343 | |||
| BC020626.1 | - | 3 | 341 | 4616168 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172575, NM_178845, AK075945, AK156529, AK145060, and others.

ZNF277
Genomic Map