siRNA Details
Gene Symbol: ZNF277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021994.2 | NP_068834.2 | 7 | 878 | ||
XM_005250129.3 | XP_005250186.1 | 6 | 680 | |||
XM_011515768.2 | XP_011514070.1 | 7 | 850 | |||
XM_017011720.1 | XP_016867209.1 | 6 | 672 | |||
GenBank mRNA | AF209198.1 | - | 7 | 878 | ||
AF308819.1 | - | 7 | 749 | |||
AK314630.1 | - | 7 | 766 | |||
AL713791.1 | - | 7 | 767 | |||
BC020626.1 | - | 7 | 765 | 4616168 |
