siRNA Details
| Gene Symbol: MSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193270.2 | NP_001180199.1 | 7 | 821 | ||
| NM_001282174.1 | NP_001269103.1 | 6 | 753 | |||
| NM_006800.3 | NP_006791.2 | 6 | 806 | |||
| NM_078628.1 | NP_523352.1 | 7 | 836 | |||
| NM_078629.3 | NP_523353.2 | 7 | 836 | |||
| XM_005274440.2 | XP_005274497.1 | 7 | 713 | |||
| XM_011545442.2 | XP_011543744.1 | 7 | 790 | |||
| GenBank mRNA | AF117065.1 | - | 7 | 836 | ||
| AK000793.1 | - | 1 | 12 | |||
| AK025642.1 | - | 6 | 780 | |||
| AK289780.1 | - | 7 | 758 | |||
| AK294255.1 | - | 6 | 623 | |||
| AK300814.1 | - | 7 | 794 | |||
| AK304419.1 | - | 6 | 634 | |||
| BC031210.1 | - | 7 | 769 | 5313214 | ||

MSL3
Genomic Map