siRNA Details
| Gene Symbol: GGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172703.2 | NP_001166174.1 | 15 | 1881 | ||
| NM_001172704.2 | NP_001166175.1 | 14 | 1782 | |||
| NM_001291641.1 | NP_001278570.1 | 16 | 2052 | |||
| NM_001291642.1 | NP_001278571.1 | 12 | 1623 | |||
| NM_014001.4 | NP_054720.1 | 14 | 1784 | |||
| NM_138619.3 | NP_619525.1 | 15 | 1883 | |||
| XM_011524563.2 | XP_011522865.1 | 15 | 1857 | |||
| XM_017024385.1 | XP_016879874.1 | 15 | 1857 | |||
| XM_017024386.1 | XP_016879875.1 | 14 | 1746 | |||
| XM_017024387.1 | XP_016879876.1 | 16 | 2006 | |||
| XM_017024388.1 | XP_016879877.1 | 16 | 2018 | |||
| XM_017024389.1 | XP_016879878.1 | 15 | 1851 | |||
| XM_017024390.1 | XP_016879879.1 | 14 | 1750 | |||
| XM_017024391.1 | XP_016879880.1 | 14 | 1736 | |||
| XM_017024392.1 | XP_016879881.1 | 14 | 1805 | |||
| GenBank mRNA | AF190864.1 | - | 14 | 1718 | ||
| AF219138.1 | - | 15 | 1818 | |||
| AF219139.1 | - | 14 | 1719 | |||
| AK124541.1 | - | 1 | 1396 | |||
| AK291651.1 | - | 15 | 1848 | |||
| AK291685.1 | - | 14 | 1715 | |||
| AK296843.1 | - | 12 | 1556 | |||
| AK301895.1 | - | 14 | 1750 | |||
| AK302278.1 | - | 15 | 1851 | |||
| AL833603.1 | - | 16 | 1992 | |||
| BC063285.1 | - | 12 | 3116 | |||
| BC070044.1 | - | 14 | 1723 | 4814878 | ||
| D63876.1 | - | 14 | 1718 | |||

GGA3
Genomic Map