siRNA Details
| Gene Symbol: CD96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318889.1 | NP_001305818.1 | 4 | 802 | ||
| NM_005816.4 | NP_005807.1 | 4 | 802 | |||
| NM_198196.2 | NP_937839.1 | 5 | 850 | |||
| NR_134917.1 | 4 | 802 | ||||
| XM_005247063.3 | XP_005247120.1 | 5 | 811 | |||
| XM_006713469.3 | XP_006713532.1 | 5 | 811 | |||
| XM_006713470.3 | XP_006713533.1 | 4 | 763 | |||
| XM_017005521.1 | XP_016861010.1 | 5 | 811 | |||
| GenBank mRNA | AK290719.1 | - | 4 | 688 | ||
| AK315237.1 | - | 4 | 684 | |||
| AL833681.1 | - | 5 | 697 | |||
| BC020749.1 | - | 4 | 733 | 4669495 | ||
| BM561433.1 | - | 4 | 802 | |||

CD96
Genomic Map