siRNA Details
| Gene Symbol: VAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199131.2 | NP_954582.1 | 4 | 1575 | ||
| GenBank mRNA | AK127095.1 | - | 4 | 1572 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VAX1| Gene Symbol: VAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199131.2 | NP_954582.1 | 4 | 1575 | ||
| GenBank mRNA | AK127095.1 | - | 4 | 1572 | ||