siRNA Details
| Gene Symbol: BCL7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197244.1 | NP_001184173.1 | 5 | 1335 | ||
| NM_001301061.1 | NP_001287990.1 | 7 | 1628 | |||
| NM_001707.3 | NP_001698.2 | 6 | 1506 | |||
| NR_036682.1 | 6 | 1489 | ||||
| GenBank mRNA | AJ223979.1 | - | 6 | 899 | ||
| BC000956.2 | - | 6 | 1465 | 3450946 | ||
| BC001967.2 | - | 6 | 1372 | 3461829 | ||
| BC009548.1 | - | 6 | 1416 | 3897805 | ||
| X89985.1 | - | 6 | 1379 | |||

BCL7B
Genomic Map