siRNA Details
| Gene Symbol: UNC5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003728.3 | NP_003719.3 | 13, 14 | 2647 | ||
| XM_005263321.3 | XP_005263378.1 | 14, 15 | 2704 | |||
| XM_017008789.1 | XP_016864278.1 | 14, 15 | 9064 | |||
| XM_017008790.1 | XP_016864279.1 | 13, 14 | 2292 | |||
| GenBank mRNA | AF055634.1 | - | 13, 14 | 2448 | ||
| AK290500.1 | - | 13, 14 | 2458 | |||

UNC5C
Genomic Map